3-Bromo-p-toluic acid ≥98%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

A11496.06 A11496.14 A11496.22
A11496.06EA 33.8 GBP
A11496.06 A11496.14 A11496.22
3-Bromo-p-toluic acid ≥98%
3-Bromo-p-toluic acid

Purity: 98+% CAS No.: 7697-26-9 Molecular Formula: C8H7BrO2 Formula Weight: 215.05 Odour: Odourless Melting Point: 206-211° Beilstein Reference: 1936510 MDL No.: MFCD00002488

Formula: C₈H₇BrO₂
Melting Pt: 206…211 °C
Storage Temperature: Ambient
MDL Number: MFCD00002488
CAS Number: 7697-26-9
EINECS: 231-712-1
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR