o-Xylene ≥99%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: 1,2-Dimethylbenzene

A11358.0F A11358.AE A11358.AP
A11358.0FEA 68.5 GBP
A11358.0F A11358.AE A11358.AP
o-Xylene ≥99%
o-Xylene

Purity: 99% CAS No.: 95-47-6 Molecular Formula: C8H10 Formula Weight: 106.17 Flash Point: 32°(89°F) Refractive Index: 1.5050 Melting Point: -25° Boiling Point: 143-145° Beilstein Reference: 1815558 Merck Reference: 14,10081 UN No.: 1307 MDL No.: MFCD00008519

Formula: C₈H₁₀
Boiling Pt: 143…145 °C
Melting Pt: –25 °C
Density: 0.877
Flash Pt: 32 °C (89 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008519
CAS Number: 95-47-6
EINECS: 202-422-2
UN: 1307
ADR: 3,III
Merck Index: 14,10081
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5dd6c62d1c62b6e7f71df7e157aae77162b3bcf1&vid=3ec8c09bb94f634e9f90b6f9e8320cd626f276d0&prid=web&clid=SAPDAM

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR