D-(-)-Tartaric acid ≥99%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

A11264.14 A11264.22 A11264.36
A11264.14EA 28 GBP
A11264.14 A11264.22 A11264.36
D-(-)-Tartaric acid ≥99%
D(-)-Tartaric acid

Purity: 99% CAS No.: 147-71-7 Molecular Formula: C4H6O6 Formula Weight: 150.09 Flash Point: 210°(410°F) Odour: Odourless pH: 1.6 Melting Point: 166-170° Beilstein Reference: 1725145 Merck Reference: 14,9068 Optical Rotation: -13° (c=10 in water) MDL No.: MFCD00004238

Formula: C₄H₆O₆
Boiling Pt: 356 °C (1013 hPa)
Melting Pt: 166…170 °C
Density: 1.76 g/cm³ (20 °C)
Flash Pt: 210 °C (410 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004238
CAS Number: 147-71-7
EINECS: 205-695-6
Merck Index: 14,9068

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR