Formamidine acetate 99

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

A11158.22 A11158.36
A11158.22EA 36.8 GBP
A11158.22 A11158.36
Formamidine acetate 99
Formamidine acetate

Purity: 99% CAS No.: 3473-63-0 Molecular Formula: C3H8N2O2 Formula Weight: 104.11 pH: 8 Melting Point: ca 158° dec. Beilstein Reference: 3563488 MDL No.: MFCD00012866

Formula: C₃H₈N₂O₂
MW: 104.11 g/mol
Melting Pt: ∼ 158 °C (dec.)
Density: 1.124
Storage Temperature: Ambient
MDL Number: MFCD00012866
CAS Number: 3473-63-0
EINECS: 222-442-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR