Phenazone ≥98%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: 2,3-Dimethyl-1-phenyl-3-pyrazolin-5-one, Antipyrine

A11089.22 A11089.36
A11089.22EA 22.7 GBP
A11089.22 A11089.36
Phenazone ≥98%
Phenazone

Purity: 98% CAS No.: 60-80-0 Molecular Formula: C11H12N2O Formula Weight: 188.23 Melting Point: 111-114° Beilstein Reference: 157775 Merck Reference: 14,716 MDL No.: MFCD00003146

Formula: C₁₁H₁₂N₂O
MW: 188.23 g/mol
Boiling Pt: 388
Melting Pt: 111…114 °C
Density: 1.25 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003146
CAS Number: 60-80-0
EINECS: 200-486-6
Merck Index: 12,00757

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR