4-Aminobutyric acid 97%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: GABA, γ-Aminobutyric acid

A11016.0E A11016.22 A11016.36
A11016.0EEA 431 GBP
A11016.0E A11016.22 A11016.36
4-Aminobutyric acid 97%
4-Aminobutyric acid

Purity: 97% CAS No.: 56-12-2 Molecular Formula: C4H9NO2 Formula Weight: 103.12 Odour: Odourless Melting Point: ca 195° dec. Beilstein Reference: 906818 Merck Reference: 14,430 MDL No.: MFCD00008226

Formula: C₄H₉NO₂
MW: 103.12 g/mol
Boiling Pt: 248
Melting Pt: ∼ 195 °C (dec.)
Density: 1.21 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008226
CAS Number: 56-12-2
EINECS: 200-258-6
Merck Index: 13,00429

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR