2-Fluorobenzoic acid ≥98%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: o-Fluorobenzoic acid

A11015.22 A11015.36
A11015.22EA 39.1 GBP
A11015.22 A11015.36
2-Fluorobenzoic acid ≥98%
2-Fluorobenzoic acid

Purity: 98+% CAS No.: 445-29-4 Molecular Formula: C7H5FO2 Formula Weight: 140.11 Melting Point: 122-125° Beilstein Reference: 971265

Formula: C₇H₅FO₂
Boiling Pt: 245 °C (1013 hPa)
Melting Pt: 122…125 °C
Density: 1.46
Storage Temperature: Ambient
MDL Number: MFCD00002405
CAS Number: 445-29-4
EINECS: 207-158-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR