Diphenylsilane 97

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

A10884.14 A10884.06 A10884.22
A10884.14EA 75.5 GBP
A10884.14 A10884.06 A10884.22
Diphenylsilane 97
Diphenylsilane

Purity: 97% CAS No.: 775-12-2 Molecular Formula: C12H12Si Formula Weight: 184.32 Flash Point: 98°(208°F) Refractive Index: 1.5790 Boiling Point: 95-97°/13mm Beilstein Reference: 2935887 MDL No.: MFCD00003002

Formula: C₁₂H₁₂Si
MW: 184.31 g/mol
Boiling Pt: 95…97 °C (13 mmHg)
Density: 0.996
Flash Pt: 98 °C (208 °F)
Storage Temperature: Ambient
MDL Number: MFCD00003002
CAS Number: 775-12-2
EINECS: 212-271-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR