Manganese(IV) oxide ≥98%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

A10765.36 A10765.0E A10765.09
A10765.36EA 58.3 GBP
A10765.36 A10765.0E A10765.09
Manganese(IV) oxide ≥98%
Manganese (IV) oxide

Purity: 98% CAS No.: 1313-13-9 Molecular Formula: MnO2 Formula Weight: 86.94 Melting Point: 535° dec. Merck Reference: 14,5730 MDL No.: MFCD00003463

Formula: MnO₂
Melting Pt: 535 °C (dec.)
Density: 5.026
Storage Temperature: Ambient
MDL Number: MFCD00003463
CAS Number: 1313-13-9
EINECS: 215-202-6
Merck Index: 14,5730

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR