3-Hydroxy-4-nitrobenzoic acid ≥98%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

A10724.06 A10724.14 A10724.22
A10724.06EA 20 GBP
A10724.06 A10724.14 A10724.22
3-Hydroxy-4-nitrobenzoic acid ≥98%
3-Hydroxy-4-nitrobenzoic acid

Purity: 98% CAS No.: 619-14-7 Molecular Formula: C7H5NO5 Formula Weight: 183.12 Melting Point: 232-236° Beilstein Reference: 2107564

Formula: C₇H₅NO₅
Melting Pt: 232…236 °C
Storage Temperature: Ambient
MDL Number: MFCD00007110
CAS Number: 619-14-7
EINECS: 210-580-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR