DL-Tartaric acid 99%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

A10683.0B A10683.30 A10683.36
A10683.0BEA 108 GBP
A10683.0B A10683.30 A10683.36
DL-Tartaric acid 99%
DL-Tartaric acid

Purity: 99% CAS No.: 133-37-9 Molecular Formula: C4H6O6 Formula Weight: 150.09 Melting Point: 210-212° dec. Beilstein Reference: 1725148 MDL No.: MFCD00071626

Formula: C₄H₆O₆
Melting Pt: 210…212 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00151254
CAS Number: 133-37-9
EINECS: 205-105-7
Merck Index: 14,9069

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR