Ammonium thiocyanate 98

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: Thiocyanic acid ammonium salt, Ammonium rhodanide, Ammonium sulphocyanate

A10632.0B A10632.0I A10632.30
A10632.0BEA 68 GBP
A10632.0B A10632.0I A10632.30
Ammonium thiocyanate 98
Ammonium thiocyanate

Purity: 98+% CAS No.: 1762-95-4 Molecular Formula: CH4N2S Formula Weight: 76.12 Odour: Odourless Melting Point: 147-149° Beilstein Reference: 3595135 Merck Reference: 14,561 MDL No.: MFCD00011428

Formula: NH₄SCN
MW: 76.12 g/mol
Melting Pt: 147…149 °C
Density: 1.305
Storage Temperature: Ambient
MDL Number: MFCD00011428
CAS Number: 1762-95-4
EINECS: 217-175-6
Merck Index: 13,00564

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR