3-Bromo-2,5-dichlorothiophene ≥97%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

A10603.03 A10603.06 A10603.14
A10603.03EA 16.7 GBP
A10603.03 A10603.06 A10603.14
3-Bromo-2,5-dichlorothiophene ≥97%
3-Bromo-2,5-dichlorothiophene

Purity: 97% CAS No.: 60404-18-4 Molecular Formula: C4HBrCl2S Formula Weight: 231.93 Refractive Index: 1.6080 Boiling Point: 95-97°/14mm MDL No.: MFCD00041440

Formula: C₄HBrCl₂S
MW: 231.93 g/mol
Boiling Pt: 95…97 °C (14 mmHg)
Storage Temperature: Ambient
MDL Number: MFCD00041440
CAS Number: 60404-18-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR