Tetraphenylphosphonium chloride ≥98%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

A10575.18 A10575.04 A10575.09
A10575.18EA 199 GBP
A10575.18 A10575.04 A10575.09
Tetraphenylphosphonium chloride ≥98%
Tetraphenylphosphonium chloride

Purity: 98% CAS No.: 2001-45-8 Molecular Formula: (C6H5)4PCl Formula Weight: 374.86 Melting Point: 272-278° Beilstein Reference: 3922393

Formula: C₂₄H₂₀ClP
Melting Pt: 272…278 °C
Storage Temperature: Ambient
MDL Number: MFCD00011916
CAS Number: 2001-45-8
EINECS: 217-890-3

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR