Sodium bromide, Wasser_1 99+% Dry weight

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

A10552.0B A10552.0I A10552.30
A10552.0BEA 47.9 GBP
A10552.0B A10552.0I A10552.30
Sodium bromide, Wasser_1 99+% Dry weight
Sodium bromide

Purity: 99+% CAS No.: 7647-15-6 Molecular Formula: NaBr Formula Weight: 102.89 Refractive Index: 1.6412 Melting Point: 755° Boiling Point: 1390° Merck Reference: 14,8594 MDL No.: MFCD00003475

Formula: NaBr
MW: 102.89 g/mol
Boiling Pt: 1390 °C
Melting Pt: 755 °C
Density: 3.203
Storage Temperature: Ambient
MDL Number: MFCD00003475
CAS Number: 7647-15-6
EINECS: 231-599-9
Merck Index: 14,8594

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR