Lithium sulphate monohydrate 99

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

A10410.0E A10410.22 A10410.36
A10410.0EEA 419 GBP
A10410.0E A10410.22 A10410.36
Lithium sulphate monohydrate 99
Lithium sulphate monohydrate

Purity: 99% CAS No.: 10102-25-7 Molecular Formula: Li2SO4.H2O Formula Weight: 127.96 (109.94anhy) Odour: Odourless Melting Point: 130° -H2O MDL No.: MFCD00149766

Formula: Li₂SO₄·H₂O
MW: 127.96 g/mol
Melting Pt: 130 °C (-H₂O)
Density: 2.06
Storage Temperature: Ambient
MDL Number: MFCD00149766
CAS Number: 10102-25-7
EINECS: 233-820-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR