Tetraethylammonium tetrafluoroborate ≥99%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: Tetrylammonium tetrafluoroborate

A10211.06 A10211.18 A10211.22 A10211.30
A10211.06EA 20 GBP
A10211.06 A10211.18 A10211.22 A10211.30
Tetraethylammonium tetrafluoroborate ≥99%
Tetraethylammonium tetrafluoroborate

Purity: 99% CAS No.: 429-06-1 Molecular Formula: C8H20BF4N Formula Weight: 217.06 Melting Point: 370-380° dec. Beilstein Reference: 3917638 UN No.: 2811 MDL No.: MFCD00011827

Formula: C₈H₂₀BF₄N
Melting Pt: 370…380 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00011827
CAS Number: 429-06-1
EINECS: 207-055-1
UN: 2811
ADR: 6.1,III

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR