Cerium(IV) sulphate hydrate ≥98%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

A10142.0B A10142.18 A10142.30 A10142.99
A10142.0BEA 638 GBP
A10142.0B A10142.18 A10142.30 A10142.99
Cerium(IV) sulphate hydrate ≥98%
Cerium (IV) sulphate hydrate

Purity: 98% CAS No.: 95838-16-7 Molecular Formula: Ce(SO4)2.xH2O Formula Weight: 332.24(anhy) Melting Point: 195° dec. Merck Reference: 14,1990 MDL No.: MFCD00149427

Formula: CeO₈S₂·H₂O
Melting Pt: 195 °C (dec.)
Density: 3.01
Storage Temperature: Ambient
MDL Number: MFCD00149427
CAS Number: 95838-16-7
EINECS: 237-029-5
ADR: ,
Merck Index: 14,1990

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR