Phenol (equilibrated) : Chloroform : iso-Amyl alcohol (25:24:1)% vol. stabilised

Supplier: PanReac AppliChem
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger


Stabilisation: Stabilised with 8-hydroxyquinoline

A0889.0100 A0889.0500
A0889.0100EA 83.4 GBP
A0889.0100 A0889.0500
Phenol (equilibrated) : Chloroform : iso-Amyl alcohol (25:24:1)% vol. stabilised
Phenol (equilibrated) : Chloroform : iso-Amyl alcohol (25:24:1 v:v:v)
Storage Temperature: Ambient
UN: 2810
ADR: 6.1,II

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR