VWR®, Low Temperature viscosity standards
Supplier: VWR Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
99478.260EA 110 GBP
99478.260 99479.260 99480.260 99481.260 99482.260 99483.260 99485.260 99486.260 99487.260 99488.260
VWR®, Low Temperature viscosity standards
Standards Viscometry Standards
Low Temperature viscosity standards have certified data for kinematic viscosity, dynamic viscosity and density for various low temperatures.
  • Fully traceable to national standards
  • Ensures full compliance to ASTM and IP test method protocols
  • Tamper-evident security packaging
Low Temperature viscosity standards are for the calibration and verification of glass capillary viscometers and other viscosity measuring equipment where operating temperature is controlled precisely.
Certifications: Certified according to ISO 17025 / ISO 17034 under UKAS accreditation.
Caution: The viscosity value given in the ordering table below is the nominal viscosity value - actual batch values are given in the certification documents supplied with the product and also on the label.
Order Now
SPECIFICATIONS
Environmentally Preferable