VWR®, Multi-Parameter Refractive Index and Density Certified Reference Material At 15, 20 and 25 °C

Supplier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

99173.030EA 324 GBP
99173.030 99174.030 99175.030 99176.030
VWR®, Multi-Parameter Refractive Index and Density Certified Reference Material At 15, 20 and 25 °C
Standards Refractive Index Standards

The Refractive Index and Density Certified Reference Materials provide a flexible, cost-effective and efficient solution to any laboratory calibrating and/or verifying refractive index and density measurements – all from a single material.


  • Density certified in accordance with primary level ASTM D1480 methodology
  • Ensures full compliance to ASTM and IP test method protocols
  • Low levels of uncertainty, ensuring maximum accuracy of data and dependable results
  • Supplied in 30 ml sealed glass vials for ample measurement readings
  • 12 month shelf life


In particular, Refractive Index and Density Certified Reference Materials are of significance to technicians with combined density and refractive index instruments to calibrate/verify their equipment’s performance and help laboratory managers meet their quality assurance requirements.


Certifications: UKAS ISO 17025 & ISO 17034.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR