J.T.Baker® Cycle-Tap, Cycle-Tap Dispenser System
Supplier: Avantor
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
9814-31EA 0 GBP
9814-31 9814-07 9814-08 9814-09 9814-10 9814-11 9814-12 9814-13 9814-14 9814-30 9814-04 9814-05 9814-06 9814-15 9814-16 9814-28 9814-29 9814-17 9814-18 9814-19 9814-20 9814-21 9814-22 9814-23 9814-24 9814-25 9814-26 9814-27 9814-32 9814-33
J.T.Baker® Cycle-Tap, Cycle-Tap Dispenser System
Bottle Top Dispensers
The cycle-tap dispenser is a stainless steel solvent dispense unit with a hose designed for unmetered manual dispensing of solvent.
  • Fully assembled with colour coded quick-connect
  • Three-way valve for purging to ensure highest purity
  • Stainless steel components that are inert to solvents to maintain solvent quality
  • Safety armored, flexible hose with internal steel braiding that will not kink or puncture
  • Thumb operated toggle valve with 'hold-on' option for easy dispensing
  • Integrated hook for hanging the dispenser’s outlet