J.T.Baker®, Liquid Level Indicators
Supplier: Avantor
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
9803-01EA 0 GBP
9803-01 9803-03 9803-00
J.T.Baker®, Liquid Level Indicators
Level Sensors
These level indicators provide the digital readout of the approximate volume of solvent remaining in the CYCLE-TAINER system.
  • Signal can also be used to drive switching circuits