VWR® LA8080, Amino acid analyser
Supplier: VWR Collection
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
939-0001EA 77090 GBP
939-0001
VWR® LA8080, Amino acid analyser
Analysers Amino Acid Analysers
The LA8080 amino acid analyser by VWR®/Hitachi combines the robustness and quality of Hitachi LC products with the performance of a post-column ninhydrin application. These instruments are used for a wide range of purposes - from measuring umami components of foodstuffs or the active ingredients in health supplements, to quality control for biopharmaceutical products. Based on components of Hitachi HPLC instruments the new bench top design enables gives scientists new possibilities in their analysis workflow. Moreover, the new design occupies 30% less surface area than its predecessors.