Supelco® TraceCERT® Organic Reference Standard, Epoxiconazole
Supplier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
93871-50MG
93871-50MGEA 108 GBP
93871-50MG
Supelco® TraceCERT® Organic Reference Standard, Epoxiconazole
Standards Organic Standards
Certifications: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI e.g. NIST or NMIJ. Certified content by quantitative NMR including uncertainty and expiry date are given on the certificate.
Caution: P202 - P273 - P280 - P308 + P313 - P391 - P405