Ethyl acetate ≥99.6% (by GC, corrected for water content), BAKER ANALYZED® HPLC for HPLC, UHPLC, for spectrophotometry, J.T.Baker®

Supplier: Avantor
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: Acetic acid ethyl ester

Packaged under nitrogen.

9282-03EA 290 GBP
9282-03 9282-02
Ethyl acetate ≥99.6% (by GC, corrected for water content), BAKER ANALYZED® HPLC for HPLC, UHPLC, for spectrophotometry, J.T.Baker®
Ethyl acetate

  • UV cutoff 255 nm

Formula: CH₃COOC₂H₅
Boiling Pt: 77.1 °C (1013 hPa)
Melting Pt: –83 °C
Density: 0.902 g/cm³ (20 °C)
Flash Pt: –4 °C
Storage Temperature: Ambient
MDL Number: MFCD00009171
CAS Number: 141-78-6
EINECS: 205-500-4
UN: 1173
ADR: 3,II
REACH: 01-2119475103-46
Merck Index: 13,03792
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=226aecffe42ffab125597552eff0ef7f88ef9983&vid=f5707ce0faee09ca10700419252c5841e0fbc01a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use = 0.01
Ultraviolet Absorbance (1.00-cm cell vs. water) - 400-330 nm ≤0.01
Ultraviolet Absorbance (1.00-cm cell vs. water) - 280 nm ≤0.02
Ultraviolet Absorbance (1.00-cm cell vs. water) - 265 nm ≤0.05
Ultraviolet Absorbance (1.00-cm cell vs. water) - UV Cut-off, nm ≤255
Fluorescence Trace Impurities, measured as Quinine Base - at 450 nm Emission, For Information Only For Information Only
Maximum for Impurities, For Information Only For Information Only
Fluorescence Trace Impurities, measured as Quinine Base - at Emission = 99.6 %
Assay (by GC, corrected for water) ≥99.6 %
Residue after Evaporation ≤2.0 ppm
Titrable Acid (µeq/g) ≤0.8
Water (by KF, volumetric) ≤0.04 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR