Ethyl acetate NF, J.T.Baker®

Supplier: Avantor
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: Acetic acid ethyl ester

9278-07EA 717 GBP
9278-07
Ethyl acetate NF, J.T.Baker®
Ethyl acetate
Formula: CH₃COOC₂H₅
MW: 88.11 g/mol
Boiling Pt: 77.1
Melting Pt: -83
Flash Pt: -4
MDL Number: MFCD00009171
CAS Number: 141-78-6
UN: 1173
ADR: 3 II
Merck Index: 13,03792
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=226aecffe42ffab125597552eff0ef7f88ef9983&vid=f5707ce0faee09ca10700419252c5841e0fbc01a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

GMP Manufactured Product
Meets N.F. Requirements
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical 99.0 - 100.5 %
NF - Assay (CH₃COOC₂H₅) 98.0 - 102.0 %
NF - Identification Passes Test
NF - Specific Gravity at 25°/25°C 0.894 - 0.898
NF - Acidity Passes Test
NF - Readily Carbonizable Substances Passes Test
NF - Nonvolatile Residue ≤0.02 %
Chromatographic Purity: Acetaldehyde ≤0.1 %
Chromatographic Purity: Ethyl isobutyl ether ≤0.10 %
Chromatographic Purity: Methyl compounds ≤0.1 %
Chromatographic Purity: Other impurities ≤0.30 %
Only Class 3 Solvents are likely to be present. Residual concentration of all Class 3 Solvents is <0.5%.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR