Acetone 99,4 GC, corrected for water content, ULTRA RESI-ANALYZED® Organic_Residue_Analysis, J.T.Baker®

Supplier: Avantor
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: 2-Propanone, DMK, Propanone, Dimethyl ketone

Inert_Gas_filled

9254.1000EA 114 GBP
9254.1000 9254.2500 9254-03
Acetone 99,4 GC, corrected for water content, ULTRA RESI-ANALYZED® Organic_Residue_Analysis, J.T.Baker®
Acetone

BAKER ULTRA RESI-ANALYZED® Reagent—ultra high-purity solvents and salts developed for organic residue extraction/concentration procedures.


These solvents are fully characterised and lot controlled by ECD, FID, or other method-specific detectors. Advanced stabiliser systems are incorporated to ensure solvent stability and superior performance. Solvents are suitable for exacting EPA sample cleanup protocols.

Formula: CH₃COCH₃
MW: 58.08 g/mol
Boiling Pt: 56.2
Melting Pt: –95.4 °C
Density: 0.792 g/cm³ (20 °C)
Flash Pt: <–20 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00008765
CAS Number: 67-64-1
EINECS: 200-662-2
UN: 1090
ADR: 3 II
REACH: 01-2119471330-49
Merck Index: 13,00067
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3de5d8e3bf7a94692d684c8f0ccdfb8500fde76d&vid=24fb2e0ec86974b1803780c7d5b23804a085df64&prid=web&clid=SAPDAM

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use Min. 99.4 %
Assay ((CH₃)₂CO) (by GC, corrected for water) ≥99.4 %
Color (APHA) ≤10
Residue after Evaporation ≤1.0 ppm
Substances Reducing Permanganate Passes Test
Titrable Acid (µeq/g) ≤0.3
Titrable Base (µeq/g) ≤0.6
Water (H₂O) ≤0.5 %
FID-Sensitive Impurities (as 2-Octanol) Single Impurity Peak (ng/mL) ≤5
ECD Sensitive Impurities (as Heptachlor Epoxide) Single Peak (pg/mL) ≤10
MEETS SPECIFICATIONS WITHIN THE EXPIRATION PERIOD


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR