Dimethyl sulphoxide 99,9 GC, corrected for water content, BAKER ANALYZED® ACS, J.T.Baker®

Supplier: Avantor
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: Methanesulphinylmethane, Methyl sulphoxide, Dimethyl(oxido)sulphur, DMSO

9224-01EA 264 GBP
9224-01
Dimethyl sulphoxide 99,9 GC, corrected for water content, BAKER ANALYZED® ACS, J.T.Baker®
Dimethyl sulphoxide

The quality of many BAKER ANALYZED™ Reagents meets or exceeds the requirements set forth by the American Chemical Society (ACS). These solvents are essential for any chemical stockroom.

Formula: (CH₃)₂SO
MW: 78.14 g/mol
Boiling Pt: 189
Melting Pt: 18.5
Density: 1.101 g/cm³ (20 °C)
Flash Pt: 87
Storage Temperature: Ambient
MDL Number: MFCD00002089
CAS Number: 67-68-5
EINECS: 200-664-3
Merck Index: 13,03285
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3ec1573e80c11ddf9254283ef76abf5fb6539c7e&vid=157e2a24c93041cbbb69833d226195e333b07665&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
Appearance Passes Test
Assay ((CH₃)₂SO) (by GC, corrected for water) ≥99.9 %
Residue after Evaporation ≤0.0100 %
Titrable Acid (meq/g) ≤0.001
Water (by KF, coulometric) ≤0.1 %
Product Information (not specifications):
Density (g/mL) at 25°C (typical) = 1.092
Meets Reagent Specifications for testing USP/NF monographs


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR