Rapid antibody labelling kits, VivoBrite™
Supplier: Biotium
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
92160 92161 92162
92160.EA 519 GBP
92160. 92161. 92162.
Rapid antibody labelling kits, VivoBrite™
Stains In vivo Imaging Dyes
These antibody labelling kits include amine-reactive near-IR CF™ dye as well as all components required for antibody labelling and purification.
Suitable for small animal in vivo imaging.
Ordering information: Each kit is sufficient for three labellings.
Caution: This item is temperature sensitive and has specific storage requirements.