VWR®, Total Nitrogen Standards (CRMs)
Supplier: VWR Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
91273.180EA 74 GBP
91273.180 91274.180
VWR®, Total Nitrogen Standards (CRMs)
Standards Environmental Analysis Standards
Certified reference materials for use as standards for Total Nitrogen, typically used as standards for Kjeldahl nitrogen determination.
  • Standard solutions for photometric applications acc EN 25663 (ISO 5663) and ISO 7150
  • Traceable to SRM from NIST
Shelf life: 24 months (12 months after opening)
Certifications: Certified Reference Material acc ISO 17025 / ISO 17034