VWR® PESTINORM®, LC Multiresidue Pesticide Standard (Mix 10) - 1 component, CRM

Supplier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

91132.001EA 40.7 GBP
91132.001
VWR® PESTINORM®, LC Multiresidue Pesticide Standard (Mix 10) - 1 component, CRM
Standards Organic Standards

A standard, typically used in food testing, Carbendazim [CAS:10605-21-7] 100 ug/ml in Methanol


All mixes are supplied with detailed certificate of analysis, chromatogram and 100% shelf-life. The CoAs provide full traceability of preparation, including the dissolution of the raw materials (with their purity), the preparation of the intermediate solutions and the preparation of the final solutions.


Certifications: Certified Reference Material (CRM) acc. ISO 17025 / ISO 17034


Caution: Storage temperature: Freezer (–20 °C)

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR