Biological indicators, self-contained tests, STERILtest
Supplier: LIOFILCHEM
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
91100 91101 20199 91150
91100.EA 598 GBP
91100. 91101. 20199-VWR 91150.
Biological indicators, self-contained tests, STERILtest
Sterilisation Indicators
Biological indicators for validating the sterilisation using steam sterilisation (autoclaves).
Devices with nutrient medium, pH indicator and discs with spores of Geobacillus stearothermophilus (GST) for steam sterilisation or Bacillus atropheus (BAT) for ethylene oxide sterilisation.