2-Propanol 99,7, BAKER ANALYZED® HPLC HPLC/UHPLC, Spectrophotometry, J.T.Baker®

Supplier: Avantor
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: Isopropyl rubbing alcohol, Isopropanol, 2-Hydroxy propane, IPA, Dimethyl carbinol, Isopropyl alcohol, Propan-2-ol

9095.1000EA 76.1 GBP
9095.1000 9095.5000 9095.2500
2-Propanol 99,7, BAKER ANALYZED® HPLC HPLC/UHPLC, Spectrophotometry, J.T.Baker®
2-Propanol
Formula: (CH₃)₂CHOH
MW: 60.1 g/mol
Boiling Pt: 82
Melting Pt: –89 °C
Density: 0.786 g/cm³ (20 °C)
Flash Pt: 12 (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00011674
CAS Number: 67-63-0
EINECS: 200-661-7
UN: 1219
ADR: 3 II
REACH: 01-2119457558-25
Merck Index: 13,05228
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ceac541820c6046c770c320cb35e80c1343901a5&vid=4a720b44a08c22b968be82ae24c2b3d604bdd52b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use min. 99.7 %
Ultraviolet Absorbance (1.00-cm cell vs. water) - 350 nm ≤0.01
Ultraviolet Absorbance (1.00-cm cell vs. water) - 280 nm ≤0.01
Ultraviolet Absorbance (1.00-cm cell vs. water) - 254 nm ≤0.020
Ultraviolet Absorbance (1.00-cm cell vs. water) - 225 nm ≤0.16
Ultraviolet Absorbance (1.00-cm cell vs. water) - UV Cut-off, nm ≤205
Assay (CH₃CHOHCH₃) ≥99.7 %
Residue after Evaporation ≤2.0 ppm
Water (by KF, coulometric) ≤0.03 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR