Software for Analytical Methods Lifecycle Assessment, NeoLiCy®
Supplier: COLLECTION PREFIX HPLC
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
NLC-M1 NLC-MX NLC-PKG-S1 NLC-S1 NLC-SMA-M1 NLC-SMA-MX NLC-SMA-S1 NLC-UP1-M1 NLC-UP1-MX NLC-UP1-S1
908-0001EA 3230 GBP
908-0001 908-0002 908-0003 908-0004 908-0005 908-0006 908-0007 908-0011 908-0012 908-0013
Software for Analytical Methods Lifecycle Assessment, NeoLiCy®
Software Chromatography Software
When operating according to quality management systems (e.g. according to GxP or ISO guidelines), statistical assessment of the analytical procedures used is very important. This includes for instance validation of analytical methods, analytical stability studies, method robustness studies, estimation of measurement uncertainty, method transfer and many other assessment steps during the whole life cycle of the analytical method. NeoLiCy® software project is designed to fulfill the requirements of all these steps according to proven and established recommendations and regulations.