Computers for Laboratory Equipment
Supplier: COLLECTION PREFIX HPLC
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
LSMPDE20TFTW LSMPDE24TFTW LSMPDE22TFTW LSAKDEBE LSAKDEIT LSAKDECH LSAKDEDE LSAKDEDK LSAKDEES LSAKDEFI LSAKDEINT LSAKDENL LSAKDENO LSAKDEPT LSAKDESE LSAKDEUK LSAKDEFR LSDEBASICW10 LSDELLLAPTOP LSDE1AIO23LTSC LSDE1W10LTSC LSDE1W10LTSCX LSDE2W10LTSC LSDE2W10LTSCX LS3W10LTSCAIC LSDE3MTLTSC LSDELLKM5221W LSPHL4202DW LSMPDE27TFTW LSPHD8132E LS3W11LTSCAIC LSDE1AIOW11 LSDE1W11LTSCX LSDE2W11LTSCX LSLTW11LTSC
906-0225EA 190 GBP
906-0225 906-0227 906-0226 906-0221 906-0212 906-0214 906-0219 906-0215 906-0218 906-0222 906-0224 906-0220 906-0223 906-0217 906-0213 906-0216 906-0211 906-0246 906-0241 906-0234 906-0236 906-0235 906-0238 906-0237 906-0240 906-0239 906-0254 906-0258 906-0253 906-0257 906-0251 906-0247 906-0249 906-0250 906-0248
Computers for Laboratory Equipment
Computer Monitors
PC equipment optimised for use with laboratory devices and software. Designed to meet laboratory conditions, ensuring necessary performance. Includes a wide range of accessories such as monitors and keyboards.
  • Pre-installed with Windows LTSC 10 or 11 to reduce downtime from frequent updates and increase stability
  • Compact footprint with integrated PC-monitor solutions
  • Pre-installed language packs
  • Keyboards with all common European language layouts
  • Compatible with main CDS (Chromatography Data Systems)