VWR® ELSD LC, ELSD FP, ELSD 100, Low Temperature Evaporative Light-Scattering Detectors
Supplier: VWR Collection
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
903-0282EA 0 GBP
903-0282 903-0283 903-0630 903-0631 903-0632
VWR® ELSD LC, ELSD FP, ELSD 100, Low Temperature Evaporative Light-Scattering Detectors
Chromatography Detectors
These ELSD models combine simplicity, reliability and robustness for all your analyses or purifications. They allow for the detection of essentially all compounds. Detection is based on a universal property of all analytes, and does not require the presence of a chromophoric group, electro-active group, etc. Control of the system can be done either locally or via a PC. A remote shutdown mode is also provided to minimise cost and enhance system lifetime.
  • Minimised band broadening thanks to a dedicated nebuliser and innovative cell design
  • SAGA (Automated
More Product Information