5(6)-Carboxyfluorescein diacetate NHS ester [CFSE (5(6)-CFDA SE)] ≥98% (by TLC) fluorescent pH indicator

Supplier: Biotium
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

90041
90041.EA 265 GBP
90041.
5(6)-Carboxyfluorescein diacetate NHS ester [CFSE (5(6)-CFDA SE)] ≥98% (by TLC) fluorescent pH indicator
5(6)-Carboxyfluorescein diacetate NHS ester [CFSE (5(6)-CFDA SE)]

5(6)-CFDA, SE (full name: 5-(and-6)-Carboxyfluorescein diacetate, succinimidyl ester, also known as CFSE) is a useful fluorescent tracer that can passively diffuse into cells and covalently label intracellular proteins, resulting in long-term cell labeling.


  • λEx/λEm (of hydrolysis product at pH 7) = 495/519 nm
  • pKa 6.5
  • Cell permeability: Membrane permeant
  • Colors: Green
  • White solid soluble in DMSO or DMF
  • Store at -20°C and protect from light


The reagent itself is colorless and non-fluorescent but becomes brightly green fluorescent once it is hydrolyzed by intracellular esterases. The labeling has been shown to be stable to formaldehyde or gluteraldehyde fixation. Also see our CFDA SE Cell Proliferation Assay Kit (catalog no. 30050).

Formula: C₂₉H₁₉NO₁₁
Storage Temperature: Freezer
MDL Number: MFCD00037466
CAS Number: 150347-59-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a0598fcd71cd7331cde037cd822c7b48750918e9&vid=a311f4fc8deb325fc63a25d6c1e5ad9256fad2b1&prid=web&clid=SAPDAM

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR