Safety Bottle Container

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

9.20078.0001 1.40140.0001
9.20078.0001EA 151 GBP
9.20078.0001 1.40140.0001
Safety Bottle Container
Carriers Bottle Carriers/Containers
These bucket-style bottle carriers are essential safety equipment for transportation of hazardous chemicals.

  • Versions for 2,5 L and 4 L glass bottles
  • Additional bottle protection by a special developed carrier with foam inlay
  • Solvent-resistant materials
  • Leak-proof top cover
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR