Tin(II) telluride ≥99.999% (metals basis)

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

89186.09 89186.18
89186.09EA 98.6 GBP
89186.09 89186.18
Tin(II) telluride ≥99.999% (metals basis)
Tin (II) telluride

Purity: 99.999% CAS No.: 12040-02-7 Molecular Formula: SnTe Formula Weight: 246.29 Physical Form: 3-12mm Pieces Density: 6.48 Melting Point: 780° UN No.: 3288 MDL No.: MFCD00049543

Formula: SnTe
Melting Pt: 780 °C
Density: 6.48
Storage Temperature: Ambient
MDL Number: MFCD00049543
CAS Number: 12040-02-7
EINECS: 234-914-8
UN: 3288
ADR: 6.1,III

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR