Iron(II) gluconate hydrate ≥97%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: Iron(II) Gluconate Hydrate, Iron(II) D-gluconate dihydrate, Gluconic Acid Iron(II) Salt

88726.36 88726.A3
88726.36EA 45.4 GBP
88726.36 88726.A3
Iron(II) gluconate hydrate ≥97%
Iron(II) gluconate hydrate

Purity: 97% CAS No.: 22830-45-1 Molecular Formula: Fe[HOCH2(CHOH)4CO2]2.xH2O (x≈2) Formula Weight: 446.19(anhy) Physical Form: Granular Melting Point: ca 140° MDL No.: MFCD00150872

Formula: C₁₂H₂₄FeO₁₅
Melting Pt: ∼ 140 °C
Storage Temperature: Ambient
MDL Number: MFCD00150872
CAS Number: 22830-45-1
EINECS: 206-076-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR