Sodium hexafluoroarsenate ≥99%

Supplier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: Sodium hexafluoroarsenate(V)

88517.09
88517.09EA 146 GBP
88517.09
Sodium hexafluoroarsenate ≥99%
Sodium hexafluoroarsenate

Purity: 99% CAS No.: 12005-86-6 Molecular Formula: NaAsF6 Formula Weight: 211.90 Physical Form: Powder UN No.: 1557 MDL No.: MFCD00014228

Formula: AsF₆Na
MW: 211.9 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00014228
CAS Number: 12005-86-6
UN: 1557
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR