Petroleum spirit, 30…40 °C, AnalaR NORMAPUR® analytical reagent

Supplier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

88261.290EA 65.9 GBP
88261.290 88261.360 88261.460
Petroleum spirit, 30…40 °C, AnalaR NORMAPUR® analytical reagent
Petroleum spirit
Storage Temperature: Ambient
MDL Number: MFCD00081849
CAS Number: 64742-49-0
EINECS: 265-151-9
UN: 1268
ADR: 3,I

Order Now

Specification Test Results

Readily carbonisable substances Passes test
Free acid (meq/g) max. 0.00008
Bromine value max. 3
Density (20/4) 0.623 - 0.627
Boiling range (30-40°C) (Vol%) min. 90%
Aromatics (as C6H6) max. 100 ppm
Evaporation residue max. 20 ppm
Sulphur compounds (as S) max. 100 ppm
Water max. 100 ppm
Al (Aluminium) max. 0.2 ppm
Ba (Barium) max. 0.1 ppm
Ca (Calcium) max. 1 ppm
Cd (Cadmium) max. 0.1 ppm
Co (Cobalt) max. 0.1 ppm
Cr (Chromium) max. 0.03 ppm
Cu (Copper) max. 0.03 ppm
Fe (Iron) max. 0.03 ppm
K (Potassium) max. 0.3 ppm
Mg (Magnesium) max. 0.1 ppm
Mn (Manganese) max. 0.03 ppm
Na (Sodium) max. 1 ppm
Ni (Nickel) max. 0.03 ppm
Pb (Lead) max. 0.1 ppm
Sr (Strontium) max. 0.03 ppm
Zn (Zinc) max. 0.3 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR