Vanadium standard solution, 1000 mg/L V in 5% nitric acid (from V₂O₃), standard for AAS, Specpure®

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

88116.AE 88116.AP
88116.AEEA 36 GBP
88116.AE 88116.AP
Vanadium standard solution, 1000 mg/L V in 5% nitric acid (from V₂O₃), standard for AAS, Specpure®
Standards Cationic Standards Vanadium Standards

Molecular Formula: V₂O₅ in 5% HNO₃
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00011453


Certifications: Traceable to NIST

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR