Thallium standard solution, 1000 mg/L Tl in 5% nitric acid (from Tl), standard for AAS, Specpure®

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

88111.AE 88111.AP
88111.AEEA 26.2 GBP
88111.AE 88111.AP
Thallium standard solution, 1000 mg/L Tl in 5% nitric acid (from Tl), standard for AAS, Specpure®
Standards Cationic Standards Thallium Standards

Molecular Formula: Tl in 5% HNO₃
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00134063


Certifications: Traceable to NIST

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR