Gallium standard solution, 1000 mg/L Ga in 5% nitric acid with hydrochloric acid (max. 1%) (from Ga), standard for AAS, Specpure®

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

88066.AE 88066.AP
88066.AEEA 34.9 GBP
88066.AE 88066.AP
Gallium standard solution, 1000 mg/L Ga in 5% nitric acid with hydrochloric acid (max. 1%) (from Ga), standard for AAS, Specpure®
Standards Cationic Standards Gallium Standards

Molecular Formula: Ga in 5% HNO₃/tr. HCl
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00134045


Certifications: Traceable to NIST

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR