Erbium standard solution, 1000 mg/L Er in 5% nitric acid (from Er₂O₃), standard for AAS, Specpure®

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

88063.AE 88063.AP
88063.AEEA 32.9 GBP
88063.AE 88063.AP
Erbium standard solution, 1000 mg/L Er in 5% nitric acid (from Er₂O₃), standard for AAS, Specpure®
Standards Cationic Standards Erbium Standards

Molecular Formula: Er₂O₃ in 5% HNO₃
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00010987


Certifications: Traceable to NIST

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR