Mercury (II) potassium iodide solution Reag. Ph. Eur. 1071500

Supplier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

87921.180EA 109 GBP
87921.180
Mercury (II) potassium iodide solution Reag. Ph. Eur. 1071500
Nessler Reagens

Ordering information: Supplied with certificate of analysis. Normally manufactured to order. Please check with customer services.

Formula: HgI₄K₂
MDL Number: MFCD00049656
CAS Number: 7783-33-7
UN: 2024
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR