Phenylhydrazinium chloride 0,36% in hydrochloric acid 5% Reag. Ph. Eur. 1064501

Supplier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: Phenylhydrazine hydrochloride, Phenylhydrazinium chloride

87892.230EA 158 GBP
87892.230
Phenylhydrazinium chloride 0,36% in hydrochloric acid 5% Reag. Ph. Eur. 1064501
Phenylhydrazinium chloride

Ordering information: Supplied with certificate of analysis. Normally manufactured to order. Please check with customer services.

Formula: C₆H₉ClN₂
MDL Number: MFCD00012924
CAS Number: 59-88-1
EINECS: 200-444-7
UN: 1789
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e16d43c3611e20636f31c95d3974860d83c8913f&vid=c26f549d3aed1b69060ac1042fd1114ca4edb2cb&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR