Antimony(III) chloride 30% (w/v) in chloroform Reag. Ph. Eur. 1007701

Supplier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

87779.180EA 193 GBP
87779.180
Antimony(III) chloride 30% (w/v) in chloroform Reag. Ph. Eur. 1007701
Antimony (III) chloride

Ordering information: Supplied with certificate of analysis. Normally manufactured to order. Please check with customer services.

Formula: SbCl₃
MDL Number: MFCD00011212
CAS Number: 10025-91-9
UN: 1760
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR